Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safranine O 0.2% in 1% acetic acid

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Safranin TSafranin Y3,7-Diamino-2,8-dimethyl-5-phenylphenazinium chlorideSafranin OGossypimine3,7-Diamino-2,8-dimethyl-5-phenylphenazin-5-ium chlorideCI 50240Basic Red 2Cotton RedSafranin ASafranin

26069-15
101410-952EA 41.7 USD InStock
101410-952
Safranine O 0.2% in 1% acetic acid
Safranine O
Safranin O Solution in Acetic Acid is a prepared, ready-to-use, high quality staining solutions for standard staining procedures used by the Biological Staining Commission and the Armed Forces Institute of Pathology

Available in concentrations 0.1% in 0.1% acetic acid and 0.2% in 1% Acetic Acid.


Formula: C₂₀H₁₉ClN₄
MW: 350.85 g/mol
MDL Number: MFCD00011759
CAS Number: 477-73-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty