Would you like to log in again?
ChemMax® 1 Disposable Chemical Protective Coveralls, Bound Seams, Elastic Wirsts and Ankles, Attached Hood and Zipper Closure, Lakeland Industries
Supplier: Lakeland Industries
C1B428Y-4X C1B428Y-XL C1B428Y-MD C1B428Y-LG C1B428Y-2X C1B428Y-3X
76314-314CS 372.01 USD InStock
76314-314 76314-316 76314-306 76314-318 76314-308 76314-310
ChemMax® 1 Disposable Chemical Protective Coveralls, Bound Seams, Elastic Wirsts and Ankles, Attached Hood and Zipper Closure, Lakeland Industries
Disposable Garments
Disposable chemical protective coveralls are made from ChemMax® 1 polyethylene barrier fabric for protection from paint, hazardous liquids, light chemical splash, and bloodborne pathogens.
  • Attached hood
  • Elastic wrists and ankles
  • Storm flap over zipper
  • Blood-borne pathogen/viral barrier tested
These garments feature bound seams to provide an additional liquid barrier for power washing and waste water treatment applications. The attached hood, elastic wrists, and elastic ankles ensure better fit and increased protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty