Would you like to log in again?
ChemMax® 1 Disposable Chemical Protective Coveralls, Bound Seams, Attached Hood and Boots, Elastic Wrists and Zipper Closure, Lakeland Industries
Supplier: Lakeland Industries
C1B414Y-XL C1B414Y-2X C1B414Y-3X C1B414Y-LG C1B414Y-4X C1B414Y-MD
76314-328CS 409.72 USD InStock
76314-328 76314-330 76314-320 76314-332 76314-322 76314-324
ChemMax® 1 Disposable Chemical Protective Coveralls, Bound Seams, Attached Hood and Boots, Elastic Wrists and Zipper Closure, Lakeland Industries
Disposable Garments
These disposable chemical protective coveralls are made from ChemMax® 1 polyethylene barrier fabric for protection from paint, hazardous liquids, chemical splash, and bloodborne pathogens.
  • Storm flap over zipper
  • Blood-borne pathogen/viral barrier tested
These garments feature bound seams to provide additional liquid barrier for power washing and waste water treatment applications. The attached hood and boots, and elastic wrists closures ensure better fit and increased protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty