Would you like to log in again?
ChemMax® 1 Disposable Chemical Protective Coveralls, Serged Seams, Attached Hood, Elastic Wrists and Ankles, Zipper Closure, Lakeland Industries
Supplier: Lakeland Industries
C1S428Y-XL C1S428Y-LG C1S428Y-2X C1S428Y-MD C1S428Y-4X C1S428Y-3X
76314-380CS 285.37 USD InStock
76314-380 76314-382 76314-372 76314-384 76314-374 76314-376
ChemMax® 1 Disposable Chemical Protective Coveralls, Serged Seams, Attached Hood, Elastic Wrists and Ankles, Zipper Closure, Lakeland Industries
Disposable Garments
These disposable chemical protective coveralls are made from ChemMax® 1 polyethylene barrier fabric for protection from paint, hazardous liquids, and chemical splash.
  • Serged seams
  • Storm flap over zipper
  • Blood-borne pathogen/viral barrier tested
These garments provide excellent protection for power washing and waste water treatment applications. Attached hood, as well as elastic wrist and ankle closures ensure increased protection.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty