Would you like to log in again?
Interceptor® Plus Level A, Gas-Tight and Vapor-Tight Chemical Protective Encapsulated Suit with Zip Front Entry and Expanded Back for SCBA, Lakeland Industries
Supplier: Lakeland Industries
INT640B-XL INT640B-2X INT640B-MD INT640B-4X INT640B-3X INT640B-LG
76314-190CS 1739.29 USD InStock
76314-190 76314-192 76314-194 76314-196 76314-198 76314-200
Interceptor® Plus Level A, Gas-Tight and Vapor-Tight Chemical Protective Encapsulated Suit with Zip Front Entry and Expanded Back for SCBA, Lakeland Industries
Disposable Garments
Level A, chemcial protective gas-tight and vapor-tight encapsulated suits are made with Interceptor® Plus chemcial protective fabric for proven protection against a broad spectrum of toxic industrial chemicals, ammonia, and fentanyl.
  • Interceptor® Plus chemical protective film
  • Level A encapsulated suit
  • Gas-tight and vapor-tight
  • Front entry design
  • Expanded back to accommodate SCBA
  • Supported by chemical toxicity risk modeling software
Interceptor® Plus chemical protective Level A encapsulated suits provide protection from the most toxic industrial chemicals and hazardous materials. These garments feature a front entry design with an expanded back to accommodate an SCBA.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty