Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Performance Series Solid Door Lab Refrigerators with Natural Refrigerants
SHCLP-23-TS SHCLP-16-TS SHCLP-12-TS SHCLP-72-TS SHCLP-26-TS SHCLP-36-TS
76169-604EA 12971.59 USD InStock
76169-604 76169-602 76169-608 76169-600 76169-622 76169-598
VWR® Performance Series Solid Door Lab Refrigerators with Natural Refrigerants
Refrigerators
The VWR® Performance Series Solid Door Laboratory Refrigerators with natural refrigerants are purpose-built for scientific and medical applications.
  • Adjustable operating temperature range: 1 - 10 °C
  • Touch screen digital temperature display and alarm module
  • Password-protected control panel
  • Sample simulated digital temperature display; 0.1 °C resolution, adjustable temperature display in Celsius or Fahrenheit
These natural refrigerant models feature new temperature displays and alarm module that offer product security with a monitoring and alarm system. Real time sample simulated temperatures are constantly displayed and supported by audible/visual high and low temperature alarms. Remote alarm dry contacts and an access port are provided to simplify connections to existing facility monitoring systems.
(Note: swivel casters add 3½" to height, which is not included in dimensions)
Certifications: ETL listed.
Ordering information: Units are supplied wth a two-year parts and labor warranty, with an additional three-year warranty on compressor parts.
Caution: (Note: swivel casters add 3½" to height, which is not included in dimensions)
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty