Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-α-Methoxyphenylacetic acid ≥99.0% (by titrimetric analysis) for gravimetric determination of Na, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: 2-Methoxy-2-phenylacetic acidDL-α-Methoxyphenylacetic acidα-Methoxyphenylacetic acid

65210-5G 65210-25G
BJ65210-5GEA 196.79 USD InStock
BJ65210-5G BJ65210-25G
(±)-α-Methoxyphenylacetic acid ≥99.0% (by titrimetric analysis) for gravimetric determination of Na, Fluka™
(±)-α-Methoxyphenylacetic acid


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 165 °C (24 hPa)
Melting Pt: 69-71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064217
CAS Number: 7021-09-2
EINECS: 230-300-9
Order Now
Specification Test Results
Identity (NMR) complying
Assay 99.0 - 101.1 %
Residue on ignition Max. 0.05 %
Melting point 69 - 71 °C
APHA Max. 50

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty