Would you like to log in again?
Determination of Elementary Electric Charge, Leybold Products
Supplier: Leybold
C1.1.3.5
470311-224EA 6240.29 USD InStock
470311-224
Determination of Elementary Electric Charge, Leybold Products
Educational Materials Chemistry Educational Materials Electrochemistry Learning Activities
Leybold Chemistry Experiments.
  • Determination of molar mass
  • States of matter
  • Analysis of Rutherford scattering
  • Chemical reactions and stoichiometry
With his famous oil-drop method, R. A. Millikan succeeded in demonstrating the quantum nature of minute amounts of electricity in 1910. He caused charged oil droplets to be suspended in the vertical electric field of a plate capacitor and, on the basis of the radius r and the electric field E, determined the charge q of a suspended droplet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty