Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

BF2649 hydrochloride ≥98% (by HPLC)

Warning
Synonyms: 1-[3-[3-(4-chlorophenyl)propoxy]propyl]piperidine hydrochloridePitolisant

AB286980-50MG AB286980-10MG
76317-776EA 275.77 USD InStock
76317-776 76317-778
BF2649 hydrochloride ≥98% (by HPLC)
BF2649 hydrochloride
BF2649 is a potent, cell-permeable and selective histamine H3 receptor inverse agonist (EC₅₀ = 1.5 nM, Ki = 0.16 nM). BF2649 displays nootropic effects in cognitive disorders; reduces locomotor hyperactivity induced by methamphetamine.

  • A potent, cell-permeable and selective histamine H3 receptor inverse agonist


Formula: C₁₇H₂₇Cl₂NO
MW: 332.31 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09970745
CAS Number: 903576-44-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty