Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

KR-33493 ≥98% (by HPLC)

Warning
Synonyms: 4-[[2-[(4-Bromophenyl)thio]acetyl]amino]-1-(2-phenylethyl)-1H-pyrazole-3-carboxylic acid

AB286998-25MG AB286998-5MG
76318-224EA 314.09 USD InStock
76318-224 76318-226
KR-33493 ≥98% (by HPLC)
KR-33493
KR-33493 is a potent FAS-associated factor 1 (FAF1) inhibitor with potential applications in treatment of Parkinson’s Disease.

  • A potent FAS-associated factor 1 (FAF1) inhibitor


Formula: C₂₀H₁₈BrN₃O₃S
MW: 460.35 g/mol
Storage Temperature: Freezer
CAS Number: 1021497-97-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty