Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

NMS-P118 ≥98% (by HPLC)

Synonyms: 2-[1-(4,4-difluorocyclohexyl)-4-piperidinyl]-6-fluoro-2,3-dihydro-3-oxo-1H-isoindole-4-carboxamide

AB287003-5MG AB287003-25MG
76318-030EA 275.77 USD InStock
76318-030 76318-028
NMS-P118 ≥98% (by HPLC)
NMS-P118
A potent, and selective poly(ADP-ribose) polymerase 1 (PARP1) inhibitor. It is selective for PARP1 (Kd = 9 nM) over PARP2 (Kd = 1,390 nM). NMS-P118 displays anticancer activity as single agent, as well as in combination therapies.

  • A potent, and selective poly(ADP-ribose) polymerase 1 (PARP1) inhibitor


Formula: C₂₀H₂₄F₃N₃O₂
MW: 395.42 g/mol
Storage Temperature: Freezer
CAS Number: 1262417-51-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty