Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

K02288 ≥98% (by HPLC)

Danger
Synonyms: 3-[6-Amino-5-(3,4,5-trimethoxyphenyl)pyridin-3-yl]phenol

AB287004-5MG AB287004-25MG
76317-878EA 206.84 USD InStock
76317-878 76317-876
K02288 ≥98% (by HPLC)
K02288
A potent and selective inhibitor of type I bone morphogenic protein (BMP) receptors (IC₅₀ values are 1.1, 1.8, 6.4, 34.4, 220, 302 and 321 nM for ALK2, ALK1, ALK6, ALK3, ActRIIA, ALK4 and ALK5 respectively). K02288 can prevent BMP4-induced SMAD1/5/8 pathway activation in vitro (IC₅₀ = 100 nM) without affecting TGF-β signaling.

  • Type I bone morphogenic protein (BMP) receptors inhibitors


Formula: C₂₀H₂₀N₂O₄
MW: 352.38 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26936347
CAS Number: 1431985-92-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty