Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-MK 801 Hydrogen Maleate ≥98% (by HPLC)

Supplier: BioVision
Synonyms: (5R,10S)-(-)-5-Methyl-10,11-dihydro-5H-dibenzo[a,d]cylcohepten-5,10-imine maleate

AB287006-25MG AB287006-5MG
76317-902EA 160.9 USD InStock
76317-902 76317-900
(-)-MK 801 Hydrogen Maleate ≥98% (by HPLC)
(-)-MK 801 Hydrogen Maleate
A less active enantiomer of the NMDA receptor antagonist, (+)-MK-801 (Cat. No. B2326)

  • A less active enantiomer of the NMDA receptor antagonist, (+)-MK-801 (Cat. No. B2326)


Formula: C₂₀H₁₉NO₄
MW: 337.37 g/mol
Storage Temperature: Freezer
CAS Number: 121917-57-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty