Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TAS-120 ≥98% (by HPLC)

Synonyms: 1-[(3S)-3-[4-amino-3-[2-(3,5-dimethoxyphenyl)ethynyl]-1H-pyrazolo[3,4-d]pyrimidin-1-yl]-1-pyrrolidinyl]-2-propen-1-one(S)-1-(3-(4-Amino-3-((3,5-dimethoxyphenyl)ethynyl)-1H-pyrazolo[3,4-d]pyrimidin-1-yl)pyrrolidin-1-yl)prop-2-en-1-one

AB287019-25MG AB287019-5MG
76317-996EA 360.07 USD InStock
76317-996 76317-998
TAS-120 ≥98% (by HPLC)
TAS-120
TAS-120 is a novel, potent and highly selective FGFR inhibitor that irreversibly inhibits all four FGFR subtypes at nanomolar concentrations. It blocks the phosphorylation of FGFRs and impedes downstream signaling pathways in cells. TAS-120 prevents the proliferation of human cancer cell lines with FGFR gene abnormalities but does not alter the growth of cell lines with wild-type FGFR.

  • A potent and highly selective FGFR inhibitor


Formula: C₂₂H₂₂N₆O₃
MW: 418.5 g/mol
Storage Temperature: Freezer
MDL Number: MFCD29037352
CAS Number: 1448169-71-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty