Would you like to log in again?
StemBoost™ Cardiac Cell Reprogramming Cocktail SCPF (100X), Biovision
Supplier: BioVision
AB286245-1ML AB286245-5
76318-010EA 649.21 USD InStock
76318-010 76318-008
StemBoost™ Cardiac Cell Reprogramming Cocktail SCPF (100X), Biovision
Cell Culture Supplements and Additives
  • A cardiac cell reprogramming cocktail
A convenient, sterile-filtered cocktail solution (100X, in DMSO) containing four (4) small molecules useful for reprogramming of mouse fibroblasts into cardiac cells. The four products are: SB-431542 (Cat. No. 1674), CHIR99021 (Cat. No. 1677), Parnate (Tranylcypromine Hemisulfate) (Cat. No. 1816), and Forskolin (Cat. No. 1531).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty