Would you like to log in again?
StemBoost™ Neuronal Fate Reprogramming Cocktail FICB (10X)
Supplier: BioVision
AB286246-5ML AB286246-1ML
76318-012EA 283.43 USD InStock
76318-012 76318-014
StemBoost™ Neuronal Fate Reprogramming Cocktail FICB (10X)
Cell Culture Supplements and Additives
  • A neuronal fate reprogramming cocktail
A convenient, sterile-filtered cocktail solution (10X, in DMSO) containing four (4) small molecules useful for reprogramming of mouse fibroblasts into functional neurons. The four products are: Forskolin (Cat. No. 1531), ISX9 (Cat. No. 2563), CHIR99021 (Cat. No. 1677), and I-BET151 (Cat. No. 2220).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty