Would you like to log in again?
Conductive ESD Trash Can, 44 Gallon, Transforming Technologies
Supplier: Transforming-Technologies
WBAS180
76209-970EA 411.13 USD InStock
76209-970
Conductive ESD Trash Can, 44 Gallon, Transforming Technologies
Bins Waste Bins
Conductive ESD trash cans remove the static threat of plastic receptacles that can damage sensitive electronic devices.
  • 44 gallon capacity
  • Permanently conductive; will never lose ESD properties
  • ESD symbol in yellow printed on body for easy identification
  • Typical point-to-point resistance: 10⁴ to 10⁵ ohm
  • Carbon loaded, black conductive polypropylene construction
ESD/anti-static trash cans keep charged waste materials away from sensitive electronic devices and from chemicals that could be ignited by a spark. When grounded, the cans and liners will not hold a static charge and they will contain materials safely. Ideal for cleanrooms, electronics manufacturing, inspection, quality control, and research laboratories. This large and durable trash can is able to withstand manufacturing environments, and will not dent, crush, or chip. Complies with IEC 61340-5-1.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty