Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Oil red O in isopropanol (IPA) fat and lipid stain

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Sudan Red 5BCI 26125Solvent Red 27

26503-02
101411-412EA 41.96 USD InStock
101411-412
Oil red O in isopropanol (IPA) fat and lipid stain
Oil Red O
For fat and lipid stain.

Stain results for Fat: Deep orange-red with Oil Red O, lighter with Sudan IV, Orange-yellow with Sudan II


Formula: C₂₆H₂₄N₄O
MW: 408.51 g/mol
Flash Pt: 13 °C (55.4 °F)
MDL Number: MFCD00003898
CAS Number: 1320-06-5
UN: 1219
ADR: 3,II
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty