Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Carboxy-2',7'-dichlorofluorescein ≥98% (by TLC) fluorescent pH indicator

Supplier: Biotium


51017
89139-256EA 139.86 USD InStock
89139-256
5-Carboxy-2',7'-dichlorofluorescein ≥98% (by TLC) fluorescent pH indicator
5-Carboxy-2',7'-dichlorofluorescein
This dye can be used as a low pH indicator similar to 51015, and is also useful as a general green fluorescent label. Carboxy-2′,7′-dichlorofluorescein is significantly more photo-stable than carboxyfluorescein.

  • λEx/λEm = 504/529 nm (pH 8)
  • ε = 90,000
  • Cell permeability: Membrane impermeant
  • Cellular localization: Fluid phase tracer
  • pKa 4.8
  • Colors: Green
  • Yellow solid soluble in DMSO, DMF or basic water (pH >7)
  • Store at 4°C and protect from light


Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty