Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ionomycin calcium salt ionophore

Supplier: Biotium

Warning

59007
89139-294EA 264.21 USD InStock
89139-294
Ionomycin calcium salt ionophore
Ionomycin calcium salt
Ionomycin is commonly used to modify intracellular calcium concentrations and to calibrate fluorescent calcium indicators. It is also known for its antiproliferative effects.

  • Yellowish oily solid soluble in DMSO or EtOH
  • Store desiccated at ≤ 4°C


Formula: C₄₁H₇₀CaO₉
Storage Temperature: Refrigerator
MDL Number: MFCD00151183
CAS Number: 56092-82-1
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty