Would you like to log in again?
Iron hematoxylin solution B Weigert's, Surgipath®
Supplier: Leica Biosystems
Synonyms: Weigert's stainWeigert's solution
Surgipath®
38016SS1D
75810-390EA 149.78 USD InStock
75810-390
Iron hematoxylin solution B Weigert's, Surgipath®
Iron hematoxylin
Component of the Leica Gomorri's Green or Blue Collagen Trichrome Stain Kits
  • Blue-black color
  • Will not dilute
Solutions A and B combine to stain the nuclei blue-black and will not be diluted out during subsequent staining steps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty