Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl sulfoxide ≥99.5%, Reagent Grade

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Methyl sulfoxideDMSOMethyl sulphoxide

D5879-25L D5879-5L D5879-2.5L D5879-500ML D5879-1GA D5879-100ML
BJD5879-100MLEA 109.17 USD InStock
BJD5879-100ML BJD5879-25L BJD5879-5L BJD5879-1GA BJD5879-500ML BJD5879-2.5L
Dimethyl sulfoxide ≥99.5%, Reagent Grade
Dimethyl sulphoxide


Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 28.5 %(V)
Melting Pt: 18.5 °C
Density: 1.104 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/77/31/2297731.jpg
Order Now
Specification Test Results
Appearance complying
Identity (NMR) complying
Assay (GC) Min. 99.5 %
Water (Karl Fischer) Max. 0.1 %
Vapor Density 2.7 (vs air)
Autoignition Temperature 573 °F
Vapor Pressure 0.42 mmHg ( 20 °C)
Explosion Limit 42 %, 63 °F
Refractive Index n20/D 1.479 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty