Would you like to log in again?
AlphaMop™ Rectangular Buckets, Texwipe®
Supplier: Texwipe
AlphaMop™
TX7062 TX7060 TX7061
10127-100EA 70.51 USD InStock
10127-100 10127-102 10127-104
AlphaMop™ Rectangular Buckets, Texwipe®
Buckets
The AlphaMop™ rectangular buckets are ideal for ceaning walls, ceilings and floors and for applying and removing solutions including disinfectants.
  • Made of PP
  • Available in three colors for easy identification
They are ideal for use with common disinfectants and cleaners.
Certifications: ISO Class 3 - 7 environments certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty