Would you like to log in again?
Rovers, Gratnells
Supplier: Gratnells
RSET054402 RSET034420 RSET054415 RSET034415 RSET034410 RSET014420 RSET034401 RSET014419 RSET054420 RSET014406 RSET014405 RSET054405 RSET034405 RSET014436 RSET054426 RSET014415 RSET014426 RSET034409 RSET054419 RSET014402 RSET054406 RSET054409 RSET014410 RSET054401 RSET034406 RSET034402 RSET034419 RSET034436 RSET014409 RSET034426 RSET054410 RSET014401 RSET054436
470316-570EA 860.15 USD InStock
470316-570 470316-572 470316-550 470316-530 470316-574 470316-552 470316-576 470316-554 470316-532 470316-556 470316-578 470316-534 470316-558 470316-536 470316-538 470316-580 470316-582 470316-560 470316-562 470316-540 470316-584 470316-586 470316-542 470316-564 470316-544 470316-566 470316-588 470316-546 470316-524 470316-568 470316-548 470316-526 470316-528
Rovers, Gratnells
Carts
Tough and practical.
  • Available in three bin size configurations: Deep, Extra Deep and Jumbo
  • Curved silver frame
  • 10" rubber tires for all terrains
  • Five sets of runners allow a variety of possible tray combinations
  • Strap to secure the trays and contents
  • Supplied fully assembled
The Gratnells Rover is a heavy duty metal trolley with durable rubber tires, runners to hold Gratnells trays, PowerTrays or SmartCases and a strap to secure your trays for transportation. This cart is perfect for transporting materials between labs and classrooms or for mobile Makerspaces. This has been designed to carry trays over any terrain and help you move heavy loads around with ease. Whether you need to take beanbags and bottled water to the field on sports day or a class full of iPads to an after school club the Rover is idea for transporting anything you need, even up and down stairs.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty