Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sanitary Reactor Adapters, Lower Outlet, 1.5" Beaded Pipe, Stainless Steel, Tri-Clamp, Chemglass

Supplier: Chemglass
CG-1964-B-155
76319-644EA 250.86 USD InStock
76319-644
Sanitary Reactor Adapters, Lower Outlet, 1.5" Beaded Pipe, Stainless Steel, Tri-Clamp, Chemglass
Fittings and Connectors
Beaded pipe to sanitary fitting adapter connects to the 1.5" beaded pipe side arm of CG-1968-GL zero dead space drain valve.

  • Approximate overall length of adapter is 1.75"
  • Adapter is manufactured from 316L stainless steel

NOTE: Requires a CG-1968 beaded pipe coupling.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty