Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

One-Piece Glass Reaction Vessel, 5-Neck, 50 ml, Chemglass

Supplier: Chemglass
CG-MR-50V1P
76319-654EA 765.6 USD InStock
76319-654
One-Piece Glass Reaction Vessel, 5-Neck, 50 ml, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
One-piece reaction vessel with five necks features: 19/22 Outer CN, (1) 10/18 Outer SN, (1) 14/20 Outer SN, and (2) 19/22 Outer SN's.

  • Vessel is graduated from 10 to 50 ml
  • Body outside diameter is 43 mm
  • Equivalent to Mettler Easymax®

Equivalent to Mettler EasyMax 102/402. Mettler Easymax® is a registered trademark of Mettler Toledo.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty