Would you like to log in again?
3B Scientific® Inverting Spectacles
Supplier: AMERICAN 3B SCIENTIFIC SE
3B Scientific®
1000895
470233-768EA 625.19 USD InStock
470233-768
3B Scientific® Inverting Spectacles
Educational Materials Physics Educational Materials Optics Learning Activities
Inverting spectacles complete with two fully rotating inverting prisms in a shielded spectacle frame.
  • Demonstration of the path of rays through an inverting prism
  • Drawing and writing
  • Seeing, picking up and balancing objects
  • Throwing and catching a ball
  • Pouring water
Inverting Spectacles Complete with two fully rotating inverting prisms in a shielded spectacle frame. The inverting prisms produce a lateral inversion of the rays – the world thus turns "upside down". Besides being a lucid demonstration of the functioning of an inverting prism, experiments involving inverting spectacles encourage a better understanding of the visual process and the functioning of the brain. This also enables a better understanding of the visual perception of babies. Even the apparently simplest things in life (holding and picking up objects, drawing, orientating oneself in a room) become inconceivably difficult.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty