Would you like to log in again?
3B Scientific® Induction Apparatus
Supplier: 3B Scientific
3B Scientific®
1022439
470233-822EA 1514.99 USD InStock
470233-822
3B Scientific® Induction Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Apparatus for demonstrating the induced voltage in a frame coil.
  • For tabletop, overhead or inclined demonstration
  • Foldable stand
Apparatus for demonstrating the induced voltage in a frame coil that is moved through the magnetic field produced by a magnet plate of limited area or by the rotation of a current-carrying conductor in the magnetic field of the magnet plate. By varying the speed of motion of the frame coil, the direction of motion and the number of turns in the coil, the induction law can be derived experimentally and quantitatively. The transparent design of the magnetic plate and coils means that they can be demonstrated on the overhead projector. An unfoldable support permits inclined setup.
Ordering information: This item does not include a power supply.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty