Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Spatulal and Spoon Tools
Supplier: GSC INTERNATIONAL SE
4-18351-PK
470313-332EA 10.35 USD InStock
470313-332
Spatulal and Spoon Tools
Spatulas
This nickel-plated spoon and spatula tool is 14.9 cm long.
The bowl of its spoon measures 30 mm by 22 mm and has a 2 ml capacity, while the spatula measures 35 mm by 18 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty