Would you like to log in again?
Cannula Holder, Stoelting
Supplier: Stoelting
51636
100229-744EA 516.54 USD InStock
100229-744
Cannula Holder, Stoelting
Cannulae
Cannula holder designed to hold 3.4 mm cannula heads, as manufactured by Plastics One™ for cannula sets, and DURECT for ALZET® osmotic pumps.
  • Non-intrusive for cementing cannulas in place
  • Holds the cannula vertically directly in line with the shaft of the probe holder
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty