Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 95-98%, ChemPUR® ACS

Supplier: Puritan Products, Inc.
Danger

287615 287675 287699 287634 287685 287690 287606 287695 287655
76266-624CS 448.98 USD InStock
76266-624 76999-006 76266-622 76266-620 76266-632 76266-630 76266-628 76266-618 76266-626
Sulfuric acid 95-98%, ChemPUR® ACS
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 1830
ADR: 8,II
Order Now
Specification Test Results
Appearance Free from Suspended or Insoluble Matter
Color Max. 10 APHA
Assay (w/w) 95.0 - 98.0%
Residue after Ignition Max. 5 ppm
Substances Reducing Permanganate Max. 2 ppm
Ammonium Max. 2 ppm
Chloride Max. 0.2 ppm
Nitrate Max. 0.5 ppm
Arsenic Max. 0.01 ppm
Heavy Metals Max. 1 ppm
Iron Max. 0.2 ppm
Mercury Max. 5 ppb

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty