Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™

Supplier: Honeywell Research Chemicals
Warning
Synonyms: Formic acid ammonium salt

55674-50G
BJ55674-50GEA 522.17 USD InStock
BJ55674-50G
Ammonium formate ≥99% (by anhydrous basis) eluent additive, for LC-MS, Fluka™
Ammonium formate


Formula: HCO₂NH₄
MW: 63.06 g/mol
Melting Pt: 116 °C
Density: 1.266 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Order Now
Specification Test Results
Assay (calculated on the anhydrous basis) 99.0 - 101.0 %
Water (Karl Fischer) Max. 2 %
Sulfated ash Max. 0.1 %
Aluminium (Al) Max. 1 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 1 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 5 ppm
Cadmium (Cd) Max. 1 ppm
Cobalt (Co) Max. 1 ppm
Chromium (Cr) Max. 1 ppm
Copper (Cu) Max. 1 ppm
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 5 ppm
Lithium (Li) Max. 5 ppm
Magnesium (Mg) Max. 1 ppm
Manganese (Mn) Max. 1 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 5 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 1 ppm
Zinc (Zn) Max. 1 ppm
Chloride (Cl) Max. 50 ppm
Total S (as SO₄) Max. 50 ppm
Suitability for the LC-MS complying
Appearance of the solution complying
Solution 1 M in H₂O complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty