Would you like to log in again?
Human Fibrinogen (from Plasma)
Supplier: EMD Millipore
Calbiochem®
341576-1GM 341576-100MG 341576-1GM 341576-1GM 341576-100MG
80051-618EA 527.97 USD InStock
80051-618 CA80051-616 CA80051-618 80051-616 MSPP-3415761GM
Human Fibrinogen (from Plasma)
Proteins and Peptides
Fibrinogen, is a soluble plasma glycoprotein that is synthesized in the hepatic parenchymal cells. Essential for clotting of blood, synthesized and secreted by hepatic parenchymal cells.
  • Purity: Homogeneous by SDS-PAGE
  • Clottable proteins: ≥90%
Prepared from plasma that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty