Would you like to log in again?
Test Kit, Sulfide, LaMotte®
Supplier: LaMotte
4456-01
66170-504EA 138.8 USD InStock
66170-504
Test Kit, Sulfide, LaMotte®
Environmental Test Kits
Sulfide test kit is used as a test for sulfide using the octa-slide 2 comparator.
Range and sensitivity is 0.2, 0.5, 1.0, 2.0, 5.0, 10.0, 15.0, 20.0 ppm S2-.
Delivery information: Kit includes :30 mL Sulfide Reagent A; 15 mL Sulfide Reagent B; 60 mL Sulfide Reagent C; 2× Test Tubes, 2.5-5-10mL, plastic, w/caps; 1× Pipet, 1.0 mL, plastic; 1× Sulfide Octa-Slide 2 Bar, 0.2-20.0 ppm; 1× Octa-Slide 2 Viewer
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty