Would you like to log in again?
Revco™ RDE Series –86 °C Ultra-Low Temperature Freezers with CO2 Back-up, Thermo Scientific
Supplier: Thermo Scientific
Revco®
RDE60086FARCO RDE60086FDRCO
76313-968EA 122723.66 USD InStock
76313-968 76313-958
Revco™ RDE Series –86 °C Ultra-Low Temperature Freezers with CO2 Back-up, Thermo Scientific
Freezers Ultra Low Temperature Freezers
Dependable –86 °C storage, powered by H-drive for operational savings and sustainability.
  • Low energy
  • Low peak variation
  • Fast recovery after door opening
  • Long warm up time
  • User friendly and quiet
Designed to keep your samples safe. Limit environmental impact and lower HVAC costs.
Certifications: SNAP Compliant
Delivery information: Package includes two shelves of sliding drawer racks and CO2 backup system.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty