Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VECTASTAIN® ABC-AmP Kit, for Mouse IgG, Chromogenic Western Blot Detection, Vector Laboratories
Supplier: Vector Laboratories
AK-6000
101097-552EA 276.92 USD InStock
101097-552
VECTASTAIN® ABC-AmP Kit, for Mouse IgG, Chromogenic Western Blot Detection, Vector Laboratories
Blotting Kits
The VECTASTAIN ABC-AmP is an amplified ABC alkaline phosphatase reagent for the detection of mouse or rabbit primary antibodies on nitrocellulose or PVDF membranes.
  • Amplified ABC alkaline phosphatase reagent for protein blot detection
  • Uses biotin/streptavidin technology
  • Contains highly sensitive chromogenic substrate, BCIP/NBT (indigo color)
  • Sufficient reagents to develop twenty 100 cm2 blots
The signal can be visualized using either a chemiluminescent/ chemifluorescent or a chromogenic substrate. Used with the DuoLuX® Chemiluminescent/Chemifluorescent substrate, the VECTASTAIN ABC- AmP system produces a very high and sustained light emission signal, with low background, and permanent fluorescence.
The sensitivity of VECTASTAIN ABC-AmP reagent combined with the DuoLux substrate allows for the detection of as little as 1 pg of target protein.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty