Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide activated acidic Brockmann Grade I ~150 mesh, Fluka™

Supplier: Honeywell Research Chemicals

199966-5KG 199966-1KG 199966-100G
BJ199966-5KGEA 524.05 USD InStock
BJ199966-5KG BJ199966-100G BJ199966-1KG
Aluminium oxide activated acidic Brockmann Grade I ~150 mesh, Fluka™
Aluminium oxide activated acidic


Formula: Al₂O₃
MW: 101.96 g/mol
Boiling Pt: 2.980 °C (1.013 hPa)
Melting Pt: 2.072 °C
Density: 3.960 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Order Now
Specification Test Results
Form powder
Brockmann Number (Activity) I (val.ref) complying
pH (10 % susp., 20°C) 4 - 5
Spez. surface (BET) 135 - 165 m²/g
Fe (as Fe₂O₃) Max. 0.03 %
Na (as Na₂O) Max. 0.4 %
Silicon (as SiO₂) Max. 0.03 %
Chloride (Cl) Max. 1.0 %
Grain size 0.04-0.30 mm Min. 95 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty