Would you like to log in again?
Stopcocks, Straight Bore, Glass Plugs, Capillary Arms, Chemglass
Supplier: Chemglass
CG-406-02
80074-076EA 84.6 USD InStock
80074-076
Stopcocks, Straight Bore, Glass Plugs, Capillary Arms, Chemglass
Stopcocks
Similar to the CG-400 series, but with the sidearms being made of capillary tubing having the same bore as the plug.
Delivery information: Supplied complete with metal retaining clip CG-423.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty