Would you like to log in again?
Stopcocks, Straight Bore, Schlenk, Glass Plugs, Chemglass
Supplier: Chemglass
CG-409-01
80074-082EA 103.73 USD InStock
80074-082
Stopcocks, Straight Bore, Schlenk, Glass Plugs, Chemglass
Stopcocks
This stopcock is specially designed for the repair of the sidearm on Schlenk vessels.
Stopcocks are available with a standard snelling to accept a CG-423-01 metal retaining clip. Stopcock has a heavy-duty, larger diameter sidearm which makes an exceptionally strong seal to the Schlenk vessel. The hose connection has an 8 mm O.D. and is 35 mm in length.
Ordering information: For replacement components see the CG-400 series.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty