Would you like to log in again?
Stopcocks, Straight Bore, Micro, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-425-01
80074-170EA 116.31 USD InStock
80074-170
Stopcocks, Straight Bore, Micro, PTFE Plugs, Chemglass
Stopcocks
Used in the fabrication of small scale apparatus, especially apparatus that has weight restrictions. PTFE plug has a 1:5 taper.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty