Would you like to log in again?
Stopcocks, Straight Bore, 1-Arm Capillary, PTFE Plugs, Chemglass
Supplier: Chemglass
CG-430-01
80074-194EA 104.64 USD InStock
80074-194
Stopcocks, Straight Bore, 1-Arm Capillary, PTFE Plugs, Chemglass
Stopcocks
Similar to the CG-424 series, but with one arm being made of 2 mm I.D. capillary tubing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty