Would you like to log in again?
Stopcocks, T-Bore, Pressure, Glass Plugs, Chemglass
Supplier: Chemglass
CG-459-01
80074-274EA 212.05 USD InStock
80074-274
Stopcocks, T-Bore, Pressure, Glass Plugs, Chemglass
Stopcocks
Similar to the CG-458 series, but with three stems sealed 90° apart.
Solid glass plug has a "T" shaped bore so that any two, or all three arms can be connected at one time. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty