Would you like to log in again?
Stopcocks, High Vacuum, Double Oblique Bore, Glass Plugs, Chemglass
Supplier: Chemglass
CG-468-02
80074-296EA 304.01 USD InStock
80074-296
Stopcocks, High Vacuum, Double Oblique Bore, Glass Plugs, Chemglass
Stopcocks
Solid glass high vacuum stopcock with the Chemglass constant pressure retention device.
This unique retaining system incorporates a glass plug having an extended snelling which accepts a stainless steel spring and clip. The spring keeps constant tension on the plug at all times and is easily removed and attached. Each stopcock is supplied complete with a stainless steel spring and clip. Replacement retention kit consists of one spring and clip for the plug size listed.
Similar to the CG-458 series, but with two parallel oblique bores. Plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty