Would you like to log in again?
Stopcocks, High Vacuum, Single Oblique Bore, Solid Glass Plugs, Chemglass
Supplier: Chemglass
CG-473-02
80074-318EA 242.52 USD InStock
80074-318
Stopcocks, High Vacuum, Single Oblique Bore, Solid Glass Plugs, Chemglass
Stopcocks
Solid plug, oblique bore stopcock is precision ground with corresponding numbers on barrel and plug.
Lower vacuum chamber is evacuated with a separate hole in the plug which is not in the line of flow.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty