Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Dimercapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 2,3-DimercaptopropanolDithioglycerineDimercaprolDimercapto-2,3 propanol-12,3-Dimercaptopropane-1-ol2,3-Dimercaptopropan-1-olBAL

38520-5ML 38520-25ML
BJ38520-5MLEA 236.19 USD InStock
BJ38520-5ML BJ38520-25ML
2,3-Dimercapto-1-propanol ≥98.0% (by GC) for complexometry, for spectrophotometric determination of Cu, Fluka™
2,3-Dimercapto-1-propanol


Formula: C₃H₈OS₂
MW: 124.22 g/mol
Boiling Pt: 120 °C (20 hPa)
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00004864
CAS Number: 59-52-9
EINECS: 200-433-7
UN: 2810
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/18/2358118.jpg
Order Now
Specification Test Results
Refractive Index n20/D 1.573 (lit.)
Assay (GC) Min. 98.0 Fl.%
IR spectrum complying
Refractive index (n 20/D) 1.573 - 1.575
Refractive Index n20/D 1.574

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty