Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™

Supplier: Honeywell Research Chemicals
Synonyms: Ehrlich's Reagent4-(N,N-Dimethylamino)benzaldehydep-Dimethylamino-Benzaldehydep-(Dimethylamino)benzaldehyde

39070-250G 39070-50G
BJ39070-250GEA 313.65 USD InStock
BJ39070-250G BJ39070-50G
4-(Dimethylamino)benzaldehyde ≥99.0% (by HPLC) for the determination of hydroxyproline, Fluka™
4-(Dimethylamino)benzaldehyde


Formula: (CH₃)₂NC₆H₄CHO
MW: 149.19 g/mol
Boiling Pt: 176-177 °C (23 hPa)
Melting Pt: 72-75 °C
Flash Pt: 164 °C
Storage Temperature: Ambient
MDL Number: MFCD00003381
CAS Number: 100-10-7
EINECS: 202-819-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/62/54/746254.jpg
Order Now
Specification Test Results
Assay (HPLC) Min. 99.0 %
Melting point 73 - 75 °C
Loss on drying Max. 0.5 %
Colour of the solution complying
Turbidity of solution complying
Residue on ignition (as SO₄) Max. 0.1 %
Identity (NMR) complying
Application test complying
Calcium (Ca) Max. 30 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Magnesium (Mg) Max. 5 ppm
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty