Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid 0.5 M CH3COOH (0.5N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

34255-1L
BJ34255-1LEA 301.17 USD InStock
BJ34255-1L
Acetic acid 0.5 M CH3COOH (0.5N) volumetric solution, Fluka™
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Factor 0.998 - 1.002

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty