Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid ≥98%, tested according to Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

45741-1L
BJ45741-1LEA 98.82 USD InStock
BJ45741-1L
Acetic acid ≥98%, tested according to Ph. Eur., Fluka™
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Explosion Limit 16 %, 92 °F
Identity A complying
Identity B complying
Appearance of the substance complying
Assay 99.0 - 100.5 %
Non-volatile matter Max. 0.01 %
Congealing - Freezing Point Min. 14.8 °C
Iron (Fe) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 23 ppm
Sulfate (SO₄) Max. 47 ppm
Reducing impurities complying
Residual solvents complying

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty