Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acetic acid ≥99.7% ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Methanecarboxylic acidVinegar acidEthanoic acid

695092-6X500ML
BJ695092-500MLCS 373.85 USD InStock
BJ695092-500ML
Acetic acid ≥99.7% ACS, Fluka™
Acetic acid


Formula: H₃CCOOH
MW: 60.05 g/mol
Boiling Pt: 118 °C (1.013 hPa)
Melting Pt: 16 °C
Density: 1.05 g/cm³ (20 °C)
Flash Pt: 40 °C
Storage Temperature: Ambient
MDL Number: MFCD00036152
CAS Number: 64-19-7
EINECS: 200-580-7
UN: 2789
ADR: 8,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/31/15/723115.jpg
Order Now
Specification Test Results
Appearance complying
Identity (IR) complying
Assay (GC) Min. 99.7 %
Non-volatile matter Max. 0.001 %
Titrable Base Max. 0.0004 meq/g
Iron (Fe) Max. 0.00002 %
Heavy metals (as Pb) Max. 0.00005 %
Chloride (Cl) Max. 0.0001 %
Sulfate (SO₄) Max. 0.0001 %
Acetic aNHydride (GC) Max. 0.01 %
KMnO₄ reducing substances complying
K₂Cr₂O₇ reducing matter complying
APHA Max. 10
Mixable with H₂O complying
Explosion Limit 16 %, 92 °F
Refractive Index n20/D 1.371 (lit.)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty