Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adipic acid 99.6-101.0% (dried basis), meets analytical specification of Ph. Eur., BP, E355, Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: 1,6-Hexanedioic acidHexanedioc acid

27635-25KG
BJ27635-25KGEA 356.3 USD InStock
BJ27635-25KG
Adipic acid 99.6-101.0% (dried basis), meets analytical specification of Ph. Eur., BP, E355, Fluka™
Adipic acid


Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
Boiling Pt: 265 °C (133 hPa)
Melting Pt: 151-154 °C
Density: 1.37 g/cm³ (20 °C)
Flash Pt: 196 °C
Storage Temperature: Ambient
MDL Number: MFCD00004420
CAS Number: 124-04-9
EINECS: 204-673-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/30/2300230.jpg
Order Now
Specification Test Results
Assay (calculated to the dried substance) 99.6 - 101.0 %
Loss on drying (105°C) Max. 0.2 %
Water (Karl Fischer) Max. 0.2 %
Sulfated ash Max. 20 ppm
Arsenic (As) Max. 3 ppm
Iron (Fe) Max. 5 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Heavy metals (as Pb) Max. 10 ppm
Chloride (Cl) Max. 50 ppm
Nitrate (NO₃) Max. 30 ppm
Sulfate (SO₄) Max. 500 ppm
Appearance of the solution complying
Related substances (HPLC) complying
Residual solvents complying
Ignition Residue ≤0.002% (as SO₄)
Vapor Density 5 (vs air)
Autoignition Temperature 788 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty